Would you like to log in again?
SP Bel-Art Heavy Duty Support Stand, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
H18303-0000
17683-060EA 161.87 USD InStock
17683-060
SP Bel-Art Heavy Duty Support Stand, Bel-Art Products, a part of SP
Support Stands
Polypropylene support stand with support rod is for use in the most severe corrosive conditions.
  • Polypropylene or polyethylene construction
  • Easy-to-clean smooth surface
  • Heavy base for stability
The base has smooth surface for easy cleaning
These heavy-duty support stands are made of PE.
Includes 12.7x762 mm (1/2x30") rod which screws into the center or either end of the base. Base is 2.5kg (5.5 lbs.) for greater stability.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty