Would you like to log in again?
NALGENE® Polypropylene Screw Caps, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
362150-5280 362150-0240 362150-4130 362150-4200 362150-6200 362150-0130 362150-4384 362150-6280 362150-6240 362150-0200 362150-5200 362150-1240 362150-4280 362150-0384 362150-1384 362150-0280 362150-5240 362150-7240 362150-7130 362150-1130 362150-7280 362150-1280 362150-5130 362150-6130 362150-4240 362150-6384 362150-1200 362150-7384 362150-5384 362150-7200
18060-130CS 135.36 USD InStock
18060-130 18060-152 18060-132 18060-154 18060-150 18060-138 16060-018 16211-008 18060-134 18060-156 16060-016 18060-136 16060-118 18060-120 18060-142 18060-144 18060-122 16060-020 16211-011 18060-140 18060-128 16060-024 16060-022 18060-124 18060-146 18060-126 18060-148 16060-026 28199-911 16211-010
NALGENE® Polypropylene Screw Caps, Thermo Scientific
Closures Bottle Closures
Linerless caps are for use only with NALGENE® narrow- and wide-mouth bottles and Erlenmeyer flasks
Note: Before autoclaving, set cap on top of container without engaging threads.
Autoclavable.
Ordering information: Bulk packed.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty