Would you like to log in again?
Nineteen-Series Steel Cabinet, Akro-Mils
Supplier: Akro-Mils
Nineteen
19320
18700-128EA 735 USD InStock
18700-128
Nineteen-Series Steel Cabinet, Akro-Mils
Furniture Storage Cabinets
Nineteen-Series steel storage cabinets feature a heavy-duty welded steel frame for strength; a chip-proof, durable, and powder-coated finish; built-in nesting feet for safe, stable stacking; and four keyhole slots for easy wall mounting. 
  • Rust, Corrosion, and Dent-Proof Polystyrene Drawers
  • Rear Stop Tab on Each Drawer
  • Dovetailed Dividers for Customized Storage Options
  • Nesting Feet for Safe Stacking
  • Four Keyhole Slots
The parts storage cabinets’ strong, high-impact polystyrene drawers are rust, corrosion and dent-proof.
The drawers also can be divided into as many as eight compartments with dovetailed dividers for customized storage options. A unique rear stop tab on each drawer prevents spills and lost materials.
Drawer dimensions: W33/16" x H21/16"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty