Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BRAND Pipette Tips, 5 ml, BrandTech

Supplier: BrandTech

702595 702600 702605
18900-052PK 87.8 USD InStock
18900-052 18900-054 12777-150
BRAND Pipette Tips, 5 ml, BrandTech
Pipette Tips
BRAND 5 ml pipette tips are slender (only 9.6 mm in diamter) and 160 mm long.

  • Tips fit BRAND and Finnpipette 5 ml pipettes
  • Made under cleanroom conditions
  • Box is repeatedly autoclavable for economy

All tips are produced under cleanroom conditions. Bulk tips are packaged in resealable bags. The TipBox holds 28 tips, is refillable, autoclavable at 121 °C, and made from heavy polypropylene for long life.

They are thin enough to pipette into an NS 12/21 stoppered flask. Tips fit BRAND Transferpette® and 5 ml Finnpipette® instruments.

Certifications: CE-IVD marked.

Ordering information: All unopened packages are DNA-, RNase-, ATP-, and endotoxin-free.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty