Would you like to log in again?
BRAND® Bottle Adapters for Dispensette®, seripettor® and Titrette®, BrandTech®
Supplier: BrandTech
704396 704343 704390 704378 704338 704429 704431 704375 704388 704397 704419 704424 704398 704325 704345 704328 704399 704322 704395 704391
89177-146EA 23.73 USD InStock
89177-146 18900-341 18900-220 18900-331 18900-210 18900-222 18900-212 18900-333 18900-224 18900-323 18900-214 89136-016 18900-226 18900-325 18900-216 18900-327 18900-228 18900-339 18900-218 18900-329
BRAND® Bottle Adapters for Dispensette®, seripettor® and Titrette®, BrandTech®
Fittings and Connectors
These polypropylene and fluoropolymer (ETFE and PTFE) adapters are for use with BRAND® bottletop dispensers and burettes.
  • Most common 4 L / 1 gal. bottles have 38 mm threads
  • Most common 1 L / 1 qt. bottles have 33 mm threads
  • Special 35 mm thread is only for use with irrigation saline used in microbiology analyzers
Outer thread refers to the outside of the adapter
Note: Many bottles come with a "neck finish" specification; a three digit number that specifies how the thread terminates on the bottle. For most laboratory bottles this is irrelevant when selecting a thread adapter. For example, for a 33-430 neck bottle, select an adapter to fit a 33mm thread.
Choose 45 mm for adapters that thread directly to current model BRAND® liquid handling instruments. The 33 mm outer thread nests inside a 45/33 mm adapter to fit smaller bottles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty