Would you like to log in again?
Motic B3-Series Upright Microscope
Supplier: Motic
1101000300891 1100100500592 1101001700971 1101000300911 1101000300891 1101001701451 1101001701291 1101001701291 1101000300782 1101000300522 1101001701491 1101001701491 1101000300802 1101000300802 1101001400392 1101001700971 1100100500611 1101001705791 1101001400392 1101000300872 1101000300811 1101000300911 1101000300522 1101000300811 1101000300872 1101000300782
19000-176EA 1896.14 USD InStock
19000-176 19000-054 19000-032 CA19000-046 CA19000-068 19000-454 CA19000-044 19000-092 19000-172 19000-050 CA19000-806 CA19000-804 19000-456 19000-458 CA19000-802 CA19000-050 19000-068 CA19000-054 CA19000-032 CA19000-092 19000-806 CA19000-456 CA19000-454 19000-802 19000-804 CA19000-458
Motic B3-Series Upright Microscope
Microscopes
The B3 Compound Microscope offers features required by more demanding microscope applications
The B3’s binocular head is inclined at 45° and can rotate 360° for comfortable viewing. The coaxial coarse and fine focus adjustments provide high precision (0.002mm) in focus adjustment. The focusable 1.25 N.A. Abbe condenser and Iris diaphragm with filter holder enable phase contrast and darkfield microscopy through the optional add-ons. Lighting is supplied through halogen illumination (12V/20W) with intensity control giving users full control over specimen illumination. Superb engineering and crystal clear optics ensure that the B3 will be a laboratory work-horse for years to come.
With built-in Koehler illumination and a standard Siedentopf head, the B3 is suitable for both educational and laboratory use. The standard achromatic super contrast (ASC) objectives provide superb image clarity and contrast. Upgrading to the optional plan achromatic objectives provides increased image flatness.
Ordering information: Microscope is supplied with all accessories listed above. Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty