Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Motic K-Series Stereo Microscopes
Supplier: Motic
1100200900182 1101001400723 1100200900161 1100200900101 1100200900161 1101001400932 1100200900101 1101001400711
19000-200EA 2712.55 USD InStock
CA19000-718 CA19000-716 19000-200 CA19000-200 19000-202 CA19000-720 19000-854 CA19000-202
Motic K-Series Stereo Microscopes
Microscopes
Motic’s K Series of stereo microscopes expand on the benefits of a Common Main Objective [CMO] optical system by integrating a Plan Achromat objective to merge the benefit of a long working distance (110mm) with the optical clarity of a CMO system
The K Series offers the highest modularity of Motic’s stereo microscopes. Choose from the K-400 series microscopes with a four-step magnification changer at 6x, 12x, 25x, and 50x ratios; the K-500 series microsopes with a five-step magnification changer at 6.4x, 10x, 16x, 25x, and 40x ratios; or the K-700 series microscopes that feature a zoom range at 1:5.2 ratio designed on self-centering dovetail ring mechanism, total parfocality, and absolute centering. Various contrast methods offer endless expandability of the K Series without the need for purchasing a new microscope or modification. One problem can be examined in two to three different methods with one microscope, without the extra space required for additional microscopes. The K Series is compatible with a variety of stands, including ball bearing and articulated boom stands, to give the most flexibility in operation. Finally, the large array of accessories such as the Discussion/Teaching System (DSK) or drawing attachment allow for seemingly endless upgradability. Whatever the need, this stereo microscope series can be easily configured and adapted to the application needs and working environment.
Images on the edge of the field of view appear flat instead of curved, as in other systems, which makes the K series ideal for inspection and photomicrography among other applications. Further enhancing the image's details and quality is the built-in double iris diaphragm of the optical bridge, which compensates for oversaturation of the illumination or the reflectiveness of the sample.
Ordering information: Microscope is supplied with all accessories listed above. Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty