Would you like to log in again?
Plain Jacket Oxygen Bomb Calorimeter, Parr®
Supplier: Parr Instrument Company
1341EB 1341EE
19420-034EA 14805.7 USD InStock
19420-034 19420-035
Plain Jacket Oxygen Bomb Calorimeter, Parr®
Calorimeters
Reliable calorimeter recommended for occasional and intermittent users (10-12 tests/day)
Molded double wall construction provides effective insulation. Using the supplied digital thermometer, standard deviations of uniform samples should not exceed 0.3%.
Excellent for teaching calorimetric determinations. Easy to set up and operate, no permanent connections required.
Certifications: CSA certified.
Ordering information: Supplied with Parr® 1108 oxygen bomb, oxygen filling connectors, Model 6775 digital thermometer, thermometer reading lens, 6 stainless steel combustion capsules, fuse wire, bomb head support stand, cover stand, calorific standard benzoic acid pellets (vial of 10), and the required ignition unit.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty