Would you like to log in again?
Double Valve Oxygen Bomb, Parr®
Supplier: Parr Instrument Company
1108 1825
19422-051EA 4255.27 USD InStock
19422-051 19440-020
Double Valve Oxygen Bomb, Parr®
Calorimeters
Designed to be easy to handle, easy to maintain and safe for determination of heats of combustion in environmental, energy and nutrition research
Provides rapid and straightforward combustion, simultaneously releasing up to 8 Kcal while oxidizing individual chemical elements for recovery analysis. Manufactured of columbium (niobium) stabilized steel (Carpenter No. 20 Cb-3), the 1108 bomb offers excellent resistance to generated nitric and sulfuric acids. The high strength bronze screw cap slides smoothly without seizing.
Used in Parr® Oxygen Bomb Calorimeters.
Max. charging pressure: 40 atm
Certifications: Factory tested according to ANSI/ASTM Method 144-64(1976), “Recommended Practice for Safe Use of Oxygen Combustion Bombs.”
Ordering information: Supplied with ANSI/ASTM certificate. Ignition units and the oxygen filling connection must be ordered separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty