Would you like to log in again?
CRUCIBLE VM W/O COVER
Supplier: PARR INSTRUMENT CO
VM Crucible^ 13mL. Nickel-chromium alloy crucible for determining volatile matter in coal and coke. Withstands repeated heating to 1000[degree]C without appreciable change in tare weight. Not for procedures requiring extreme corrosion resistance. No insert cover.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRUCIBLE VM W/O COVER 19422-086 3102 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty