Would you like to log in again?
HYGEN™ Microfiber Dry Room Mop, Rubbermaid®
Supplier: Rubbermaid
RCPQ412GRE
19758-829EA 33.58 USD InStock
19758-829
HYGEN™ Microfiber Dry Room Mop, Rubbermaid®
Mops
High-pile, 100% microfiber collects and holds dust and dirt mechanically and electrostatically for superior dusting results
Dense 3 - 5 micrometer quality fiber removes more dust, dirt and microbes than conventional cleaning products. Advanced hook-and-loop backing; holds-up under harsh laundering conditions and attaches pads more securely. Double finished edges retain mop shape with use over time. Color coded products help reduce cross contamination by differentiating area of use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty