Would you like to log in again?
MetroMax i™/MetroMax iQ™ Stem Casters, Metro™
Supplier: Metro International
MetroMax i® MetroMax i™
5MDRXA 5MX 5MPXGSA 5MDRXA 5MBX 5PCXM 5MPXGSA 5MBX 5MX 5MDBXA 5MRX 5MDXA 5MDBXA 5MPBX 5PCBX 5MRX 5MPRXGSA 5PCBX 5MPRX 5PCX 5MPBXGSA 5PCBXM 5PCRX 5MPX 5MPX 5PCXM 5PCX 5MPRXGSA 5PCBXM 5MPBXGSA 5MDXA 5MPRX 5MPBX 5PCRX
22233-644EA 142.56 USD InStock
22233-644 22233-642 22233-640 CA97005-872 CA97005-750 CA19766-476 CA82027-250 97005-754 CA19766-254 CA82027-252 19766-486 19766-242 CA19766-494 CA19766-252 CA19766-478 19766-248 CA22233-642 CA22233-640 CA22233-644 97005-872 97005-750 97005-752 CA97005-752 CA19766-486 82027-252 19766-478 CA19766-242 19766-476 CA97005-754 19766-254 19766-494 19766-252 82027-250 CA19766-248
MetroMax i™/MetroMax iQ™ Stem Casters, Metro™
Furniture Shelves
Use with MetroMax i™ or MetroMax iQ™ shelving to create mobile units.
  • Resilient rubber tread casters are non-marking and feature a molded, soft tread that provides quiet operation
  • High modulus rubber casters provide extra shock protection
  • Polyurethane tread casters are abrasion-resistant, non-marking, and shock-absorbent
Casters with all-polymer horn offer corrosion resistance and enhanced durability.
Each caster includes a protective bumper. Casters feature a tread width of 3.2 cm (⁵/₄") and a wheel diameter of 12.7 cm (5"). Swivel, foot-operated brake, and rigid styles are available.
Ordering information: Rigid casters are held in place by a connecting channel; when ordering rigid casters, shelf depth must be provided.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty