These utility carts are designed for long life
All 45.7cm (18") wide carts have 10.1cm (4") light-duty casters. The 53.3cm (21") and 61cm (24") wide carts have 12.7cm (5") light-duty casters. All carts include 8.9cm (31/2") diameter donut bumpers. Overall height (to top of handle) is 101cm (40").
MW100 series carts feature two stainless steel flat shelves and stainless steel handles. MW200 series carts feature three stainless steel flat shelves and stainless steel handles. MW400 series carts feature two chrome wire shelves, one stainless steel flat shelf, and chrome handles. Solid shelf can be positioned on top, middle, or bottom. MW600 series carts feature two wire shelves in chrome or stainless steel with handles of matching material. MW700 series carts feature three wire shelves in chrome or stainless steel, with handles of matching material.
Flat shelves are ideal for transporting materials that require an expanse of flat, solid surface. They feature a raised edge, which permits fast cleanup of spilled liquids, and double-thick 18-gauge steel at the edges for rigidity and strength. Wire shelves have a bright, modern, sanitary appearance. Their open construction minimizes dust accumulation and maximizes visibility and air circulation.
They are maintenance-free and easy to maneuver in small areas.