Would you like to log in again?
Perforated Top Clean Tables, Metro™
Supplier: Metro International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CTP3072S-H CTP3036S CTP3072S-H CTP3036S CTP3060S CTP3072S CTP3060S-H CTP3060S CTP3072S CTP3048S CTP3048S-H CTP3060S-H CTP3048S CTP3048S-H CTP3036S-H CTP3036S-H
19766-708EA 5042.89 USD InStock
CA19766-708 CA19766-706 19766-708 19766-698 CA19766-704 19766-706 CA19766-702 CA19766-712 19766-704 19766-712 CA19766-700 CA19766-710 19766-702 CA19766-698 19766-710 19766-700
Perforated Top Clean Tables, Metro™
Furniture Tables and Workbenches
These clean tables are made of type 304 stainless steel with HD super posts and super Erecta® construction throughout.
  • The perforated top is manufactured from 14-gauge
  • Support members have a satin finish
Type 304 stainless steel with 1.9 cm (³/₄") holes on 2.5 cm (1") centers. These units provide approximately 35% open area.
Perforated top tables are available in the overhead style with 188 cm (74") high rear posts. Use the cantilever shelves, grid panels and accessories to customize the work area.
All perforated top tables feature height adjustable legs 83.8 to 101.6 cm (33 to 40") in 2.5 cm (1") increments. Shipped knocked down, the units can be easily assembled in minutes.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty