Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Deep Ledge Utility Carts, Metro™

Supplier: Metro International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BC2636-2DBL BC2636-2DG BC2030-3DBL BC2636-3DG BC2030-2DG BC2030-3DG BC2030-3DG BC2636-2DG BC2636-3DG BC2030-2DBL BC2636-3DBL BC2030-3DMB BC2030-2DMB BC2030-2DG BC2030-3DBL BC2636-2DBL BC2636-3DBL
82021-100EA 605.75 USD InStock
82021-100 82026-662 89414-858 89414-856 19766-874 19766-850 CA19766-858 CA82026-668 CA82026-666 CA19766-866 82026-668 19766-858 CA82026-664 19766-866 CA82026-662 CA19766-874 82026-664
Deep Ledge Utility Carts, Metro™
Carts
Utility carts feature corrosion-proof polyurethane shelves with 7cm (23/4") deep ledges to contain products and spills

Smooth surfaces are designed to resist staining and wipe clean easily.

They have chrome-plated posts and an ergonomically-designed, chrome-plated handle. Carts are mounted on four resilient 10.2cm (4") diameter rubber casters. Three-shelf units have an easy-to-position center shelf that is adjustable in 2.5cm (1") increments. Each shelf can hold up to 68kg (150lbs.), and each unit can hold up to 181kg (400lbs.). Height: 104cm (41").

Certifications: NSF listed.

Ordering information: Carts are shipped knocked down and require no tools to assemble.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty