Would you like to log in again?
Super Erecta® Stem Casters, Metro™
Supplier: Metro International
Super Erecta™ Super Erecta®
5MDBA 5MFA 5MR 5MDBGSA 6MP 6MPR 5MDA 5MPR 5M 5MDRGSA 5MFBA 5MFBA 5M 5MDA 5MC 5MESD 5MBC 6MPB 5MP 6MPB 6MPR 4LD 5MDGSA 5MBESD 5LD 5MC 6MP 5MPB 5MBESD 5MPR 5MB 5MR 5MP 5MESD 5MFA 5MDBGSA 5LD 5MDRA 5MDGSA 4LD 5MDBA 5MDRA 5MB 5MBC 5MDRGSA
19767-004EA 136.08 USD InStock
19767-004 CA97005-490 19766-480 97005-486 19767-006 97005-488 19766-246 CA19766-794 CA97005-488 CA22221-200 CA19766-474 19766-244 CA19766-792 CA22221-204 CA19766-250 CA19766-492 19766-482 19766-240 97002-896 97002-894 CA19766-238 CA22221-188 CA19766-236 22221-198 97005-490 CA19766-816 19766-492 19766-250 CA22221-208 CA19767-004 CA19767-006 CA97005-486 22221-204 CA19766-244 19766-236 19766-794 19766-792 CA19766-482 CA19766-240 19766-474 CA19766-480 CA22221-198 22221-200 19766-238 22221-188
Super Erecta® Stem Casters, Metro™
Furniture Shelves
For use with Super Erecta® shelving and SiteSelect® posts to create mobile units. Swivel, foot-operated brake, and rigid styles are available.
  • Resilient rubber tread casters are non-marking and feature a molded, soft tread that provides quiet operation
  • Polyurethane tread casters are abrasion-resistant, non-marking, and shock-absorbant
Casters with high modulus rubber tread provide extra shock protection and have excellent resistance to oil and grease. Special cart-washable casters are constructed of stainless steel components. Casters include grease seals and zerk fittings to withstand high-pressure washings.
Conductive casters conform to Federal Specification FF-C-88C regarding the acceptable standards for conductive wheels. To ensure proper conductivity, aluminum split sleeves must be used with each shelf.
Ordering information: Rigid casters are held in position by a connecting channel; when ordering rigid casters, shelf width must be provided. An 8.9 cm (3¹/₂") diameter donut is furnished standard on each caster.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty