Would you like to log in again?
Super Erecta® Wire Shelving, Stainless Steel, Metro™
Supplier: Metro International
Super Erecta™ Super Erecta® MAXYMum Recovery™
3636NS 2160NS 2448NS 2142NS 1430NS 1430NS 2460NS 1848NS 1842NS 2124NS 2160NS 1842NS 2436NS 2148NS 2148NS 1860NS 2454NS 1854NS 1424NS 1836NS 1824NS 1472NS 2124NS 2448NS 1448NS 1824NS 2130NS 2472NS 1830NS 1460NS 2136NS 3648NS 3636NS 2442NS 1436NS 3660NS 3660NS 3048NS 2424NS 3036NS 1836NS 1848NS 2142NS 3672NS 2454NS 2430NS 2172NS 3060NS 2436NS 3672NS 2442NS 2172NS 2424NS 1442NS 2460NS 3072NS 1460NS 1472NS 2472NS 2136NS 1872NS 1442NS 2154NS 1854NS 1436NS 1448NS 2154NS 2130NS 1830NS 2430NS 1424NS 1872NS 1860NS
19766-084EA 409.72 USD InStock
CA19767-056 19766-084 CA19766-092 19766-522 CA19766-530 19766-168 19766-520 CA19766-098 19766-088 19766-122 CA19766-538 19766-528 CA19766-536 19766-526 CA19766-534 19766-204 19766-524 CA19766-532 CA19766-518 19766-074 19766-072 CA19766-168 CA19766-520 19766-510 CA19766-122 CA19766-088 CA19766-084 19766-518 CA19766-528 19766-516 CA19766-526 CA19766-524 CA19766-204 19766-512 CA19766-522 CA19766-508 CA19766-072 19766-140 19766-544 19766-542 19766-540 19766-386 CA19766-074 19766-508 19767-060 CA19766-516 19767-062 19766-506 CA19766-512 19766-504 CA19766-510 19767-056 19767-058 19767-052 CA19767-062 19767-054 19766-092 19766-532 CA19766-542 CA19766-540 19766-530 CA19766-386 CA19766-140 19766-098 CA19766-506 19766-538 CA19767-060 19767-050 CA19766-504 19766-536 CA19766-544 19766-534 19766-810
Super Erecta® Wire Shelving, Stainless Steel, Metro™
Furniture Shelves
Type 304 stainless steel shelves provide maximum protection in corrosive environments.
  • Open-wire design
  • Easy shelf adjustment
  • Double grooves to aid shelf alignment
Open-wire design minimizes dust accumulation, allows a free circulation of air, and provides greater visibility of stored items
Shelves up to 121.9 cm (48) in length can support up to 363 kg (800 lbs.) per level. Shelves longer than 121.9 cm (48") can support up to 272kg (600lbs.) per level.
Note: The 35.6 cm (14) wide shelves require the use of footplates (see 19766-814 series) when used as a freestanding unit. The actual length of the shelves is 3.2 mm (1/8") shorter than the nominal dimension shown. Stationary posts over 189.6 cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap.
Easy shelf adjustment is possible through the use of SiteSelect® stationary or mobile posts (sold separately), which allow shelves to be readjusted and snapped into place at precise 2.5 cm (1) increments along the entire height of the post. An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. Stem casters (see 19766-236 series) and shelf dividers and ledges (see 22233-212 series) are available separately.
Certifications: Shelves meet MIL-S-40144E specifications.
Ordering information: Each individual shelf and post is sold separately. To order posts or accessories, contact your Avantor representative for more information. Each shelf includes four pairs of plastic split sleeves for securing shelf to posts.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty