Would you like to log in again?
GLOVE VNYL PF CLS 100 M CS100
Supplier: PIP
CleanTeam* Disposable Gloves for Critical Environments^ Class 10 cleanroom vinyl, 12 inch length, powder free, 5 mil. Electrostatic dissipative. Beaded cuff. Packed 100 gloves double bagged in ESD poly bag. Doubled boxed case. Medium.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE VNYL PF CLS 100 M CS100 20000-260 100-2830/M Case of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty