Would you like to log in again?
CleanTeam® Medium Weight Lisle Inspection Gloves, Protective Industrial Products
Supplier: Protective Industrial Products
CleanTeam®
97-520H 97-520R 97-520/12R 97-520J 97-521/10 97-520/10 97-520 97-521H 97-521
32932-322CS 1991.18 USD InStock
32932-322 20000-530 20000-540 20000-544 32932-217 20000-543 32932-324 20000-535 32932-215
CleanTeam® Medium Weight Lisle Inspection Gloves, Protective Industrial Products
Gloves
Constructed in a two-piece, reversible pattern using 100% cotton fabric.
  • Ideal for inspection and electronics assembly
  • Designed to minimize contamination of products from human hands
  • Can be used as liners under disposable and unsupported liquid proof gloves
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty