Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IPTG (isopropyl-β-D-thiogalactopyranoside) animal-free

Supplier: TEKNOVA INC
Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triolIsopropyl Beta-D-1-thiogalactopyranoside

I0097
200059-640EA 143.85 USD InStock
200059-640
IPTG (isopropyl-β-D-thiogalactopyranoside) animal-free
IPTG (Isopropyl-β-D-thiogalactopyranoside)


Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty