Would you like to log in again?
EZ Rich Defined Medium Kit, Teknova
Supplier: Teknova
EZ Rich
M2105 M2105
200059-658EA 490.38 USD InStock
CA200059-658 200059-658
EZ Rich Defined Medium Kit, Teknova
Media for Microbiology Prepared Media for Microbiology
Designed for the growth of E
MOPS mixture, ACGU solution, and EZ supplement should be stored frozen. The potassium phosphate dibasic solution and 20% glucose solution should be stored at room temperature.
coli and other enterobacteria. Kit consists of 100mL 10X MOPS mixture (200059-678), 50mL 0.132M potassium phosphate diabase solution (200059-652), 100mL 10X ACGU solution (200059-654), 200mL 5X supplement EZ (200059-656), and 100mL 20% glucose solution (100216-728). Kit makes 5 liters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty