Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OPN Mouse Monoclonal Antibody [clone: 53]

Supplier: Enzo Life Sciences
905-629 905-629-100
200061-474EA 409 USD InStock
200061-474 CA200061-474
Anti-OPN Mouse Monoclonal Antibody [clone: 53]
Antibodies
Osteopontin (OPN) is an acidic extracellular matrix cell adhesion protein that is relatively abundant not only in bone matrix, plasma, urine, and milk, but is also found in malignant and atherogenic tissues. Phosphorylation, glycosylation and calcium modifications allow intact and fragmented Osteopontin to direct a variety of diverse responses including tissue remodeling, inflammation and cell survival. Plasma Osteopontin has been shown to provide prognostic information in breast, prostate, colon, and lung cancers as well as metastatic carcinomas. The notable presence of Osteopontin in a variety of tumors is strongly correlated to pathological stage, suggesting its critical role in tumor invasiveness, progression and metastasis. In addition, Osteopontin inhibits inducible nitric oxide synthase activity, thereby protecting tumor cells from NO-mediated macrophage cytotoxic attack. Osteopontin is found in atherosclerotic plaques and may drive a number of diabetic vascular pathologies.

Type: Primary
Antigen: OPN
Clonality: Monoclonal
Clone: 53
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty