Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP7 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
AAM-137E ADI-AAM-137-C ADI-AAM-137-E AAM-137C
200061-508EA 387 USD InStock
200061-508 CA200061-506 CA200061-508 200061-506
Anti-CASP7 Mouse Monoclonal Antibody
Antibodies
The caspases are a family of cysteine proteases that cleave after certain aspartate residues, and are primarily recognized as mediators of apoptosis. caspases are synthesized as inactive zymogens that can be cleaved to form active enzymes following the induction of apoptosis by stress or death receptors. Initiator caspases (e.g. caspase-8 and -10) are activated by dimerization of the zymogen on a dedicated adaptor protein. These activated initiator caspases in-turn cleave downstream effector or executioner caspases (e.g. caspase-3, -6, and -7) in a cascade-like manner, which cleave key cellular proteins that lead to the morphological changes associated with apoptotic cell death.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, hamster and monkey Caspase-7.

Type: Primary
Antigen: CASP7
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty