Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AGRN Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
AGR-530F AGR-530D ADI-AGR-530-F ADI-AGR-530-D
200061-660EA 294 USD InStock
CA200061-662 200061-660 200061-662 CA200061-660
Anti-AGRN Mouse Monoclonal Antibody
Antibodies
Agrin is an essential extracellular matrix component which promotes clustering of nicotinic acetylcholine receptors (nAChRs) and other proteins at the neuromuscular junction during development. Agrin, MuSK, and Rapsyn are all essential components for AChR aggregation, through an unknown mechanism. The C-terminal region of agrin is released into the medium, interacts with receptors on the muscle surface and induces AChR aggregation.

  • For Immunocytochemistry, Western Blot

Recognizes mouse and rat agrin. Detects a band of ~100kDa by Western blot.

Type: Primary
Antigen: AGRN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty