Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AGRN Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
AGR-540D ADI-AGR-540-F ADI-AGR-540-D AGR-540F
200061-664EA 265 USD InStock
CA200061-664 200061-664 CA200061-666 200061-666
Anti-AGRN Mouse Monoclonal Antibody
Antibodies
Agrin is an essential extracellular matrix component which promotes clustering of nicotinic acetylcholine receptors (nAChRs) and other proteins during development at the neuromuscular junction. Agrin, MuSK and Rapsyn are all essential components for AChR aggregation through an unknown mechanism. The C-terminal region of agrin is released into the medium, interacts with receptors on the muscle surface, and induces AChR aggregation. The central region contains two O-linked glycosylation sites and a domain homologous to domain III of laminin. The N-terminal region anchors agrin to the extracellular matrix via other basal membrane components. This region also contains a protease inhibitor domain and glycosaminoglycan attachment sites, increasing the predicted MW from 200kDa to ~600kDa. The diagram shown indicates the domain structure and functional regions of agrin, as well as domains required for AChR aggregation and alpha-dystroglycan and heparin binding.  Various agrin isoforms are generated by alternative splicing at the X, Y and Z sites, and differ in the presence or absence of small inserts. The isoforms can determine the biological activity of agrin and their expression in specific tissues and stages of development. While no difference in functional activity has been detected between splicing variants at site X, insertion of a 4 aa peptide at site Y modestly increases agrins nAChR clustering activity. Insertion of an 8 aa peptide at splicing site Z increases the clustering activity of soluble rat agrin 10,000 fold.

  • For Immunohistochemistry, Western Blot

Recognizes mouse and rat Agrin.

Type: Primary
Antigen: AGRN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty