Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TAP1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
CSA-620E ADI-CSA-620-E
200061-758EA 421 USD InStock
CA200061-758 200061-758
Anti-TAP1 Rabbit Polyclonal Antibody
Antibodies
TAP1 and TAP2 are two structurally related subunits of the Transporter associated with antigen processing (TAP). The TAP complex is a member of the ATP binding cassette (ABC) family of transmembrane transporters. The TAP transporter is an essential component of the MHC class I antigen presentation pathway by binding peptides in its cytosolic part and subsequently translocating the peptides into the lumen of the endoplasmic reticulum where assembly of MHC class I and pepide takes place.

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human and rat TAP1. Detects a band of ~70kDa by Western blot.

Type: Primary
Antigen: TAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty