Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TCP1A Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-CTA-191-D CTA-191F CTA-191D ADI-CTA-191-F
200063-090EA 547 USD InStock
CA200063-090 200063-090 CA200061-820 200061-820
Anti-TCP1A Rat Monoclonal Antibody
Antibodies
TCP-1 alpha is a 60 kDa subunit of a cytosolic hetero-oligomer chaperone that is known to be involved in the folding of Actin and Tubulin. This protein is a member of the Chaperonin family, which includes E. coli GroEL, the mitochondrial Heat-shock protein Hsp60, the plastid Rubisco-subunit-binding protein, and the archaebacterial protein TF55. These Chaperonins assist the folding of proteins upon ATP hydrolysis.

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, dog, C. elegans, Drosophila, guinea pig, hamster, monkey, pig, plant, rabbit and yeast TCP-1alpha. Detects a band of ~60kDa by Western blot.

Type: Primary
Antigen: TCP1A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty