Would you like to log in again?
Anti-CUL1 Mouse Monoclonal Antibody
Supplier: Enzo Life Sciences
KAM-CC135-D KAM-CC135F KAM-CC135D KAM-CC135-F
200061-860EA 273 USD InStock
CA200061-860 200061-860 CA200061-862 200061-862
Anti-CUL1 Mouse Monoclonal Antibody
Antibodies
The human Cullin family of genes consists of Cul-1, Cul-2, Cul-3, Cul-4a, Cul-4b, and Cul-5. Among the 6 members, Cul-1 and Cul-2 are very well characterized. Cul-2 is homologous to the Saccharomyces cerevisiae cdc53 gene. Studies have shown that Cul-1 forms an E3 ubiquitin-ligase, called the SCF complex (Skp1 and F-box protein such as bTrCp or FWD1), which controls the proteolysis of G1 cell cycle regulatory proteins.
  • For Western Blot
Recognizes human, mouse, rat, bovine, monkey and rabbit Cullin 1. Detects a band of ~93kDa by Western blot.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty