Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EIF4E-BP2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-MA112-E KAP-MA112E
200062-088EA 358 USD InStock
CA200062-088 200062-088
Anti-EIF4E-BP2 Rabbit Polyclonal Antibody
Antibodies
The eukaryotic translation initiation factors (eIFs) regulate translation at the level of messenger RNA engagement by the ribosome. eIF-2alpha promotes the binding of the initiator tRNA to 40S ribosomal subunits, and is a key signaling component of the unfolded protein response (UPR). eIF-4E recruits mRNAs to the eIF4F complex through binding of their 5 N7-methylguanosine cap. eIF-4E binding proteins-1 and -2 are translational repressors, sequestering eIF-4E in conditions of low cellular demand for translation. Their dissociation from eIF-4E is regulated by phosphorylation in response to growth-promoting stimuli.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, pig and rabbit eIF4E-BP2.

Type: Primary
Antigen: EIF4E-BP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty