Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PKC Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-PK003-E KAP-PK003E
200062-116EA 385 USD InStock
200062-116 CA200062-116
Anti-PKC Rabbit Polyclonal Antibody
Antibodies
Protein Kinase C (PKC) is a large superfamily of serine/threonine kinases that mediate essential cellular signals required for activation, proliferation, differentiation and survival. There are at least ten PKC isotypes that are closely related in structure but that have distinct patterns of tissue distribution and function. The PKC isotypes can be subdivided into three classes based on primary structure and biochemical properties. These are: classical or conventional PKC isotypes (cPKC), novel PKC isotypes (nPKC) and atypical PKC isotypes (aPKC).

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, monkey, pig, rabbit and sheep PKC. Detects a band of ~67-83kDa by Western blot.

Type: Primary
Antigen: PKC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty