Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Dehydrin Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PLA-100D ADI-PLA-100-F PLA-100F ADI-PLA-100-D
200062-430EA 226 USD InStock
CA200062-432 200062-430 CA200062-430 200062-432
Anti-Dehydrin Rabbit Polyclonal Antibody
Antibodies
Dehydrins are a family of proteins that become abundant during dessication in seedlings and embryos of cereal crop plants including barley, corn, wheat, and rice. Dehydrin proteins have a highly conserved lysine-rich block (KIKEKLPG) found near the carboxy terminus which appears to be characteristic of, and unique to Dehydrins.

  • For Western Blot

Recognizes plant Dehydrin. Detects a band of ~65kDa by Western blot.

Type: Primary
Antigen: DEHYDRIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Plant

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty