Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ubiquitin Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SPA-200F ADI-SPA-200-D ADI-SPA-200-F SPA-200D
200062-496EA 544.7 USD InStock
CA200062-494 200062-496 CA200062-496 200062-494
Anti-Ubiquitin Rabbit Polyclonal Antibody
Antibodies
Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by the 26S complex, a multicatalytic cytosolic and nuclear protease. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.

  • For Immunocytochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, Drosophila, fish, monkey, hamster, rabbit, guinea pig, pig, sheep, Xenopus and yeast Ubiquitin. Detects a band of ~10kDa by Western blot.

Type: Primary
Antigen: UBIQUITIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty