Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ubiquitin Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
SPA-203F ADI-SPA-203-F ADI-SPA-203-D SPA-203D
200062-504EA 576 USD InStock
CA200062-502 200062-504 CA200062-504 200062-502
Anti-Ubiquitin Mouse Monoclonal Antibody
Antibodies
Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by the 26S complex, a multicatalytic cytosolic and nuclear protease. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.

  • For Western Blot

Recognizes human, mouse, rat, bovine, hamster, and rabbit Ubiquitin. Detects a band of ~6.5kDa by Western blot.

Type: Primary
Antigen: UBIQUITIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty