Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GROES Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-210-D SPA-210F ADI-SPA-210-F SPA-210D
200062-506EA 201 USD InStock
200062-506 CA200063-248 CA200062-506 200063-248
Anti-GROES Rabbit Polyclonal Antibody
Antibodies
GroEL and GroES are E. coli chaperonins, which are homologs of mitochondrial chaperonins Hsp60 and Hsp10. GroEL is a double toriodal assembly of 14 identical subunits which form two heptameric rings stacked back-to-back, with a cavity at each end. GroEL and its co-chaperonin GroES facilitate protein folding with an ATP-dependent mechanism.

  • For Western Blot

Recognizes bacteria GroES. Detects a band of ~10kDa by Western blot.

Type: Primary
Antigen: GROES
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty