Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRYAA Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-221D ADI-SPA-221-D ADI-SPA-221-F SPA-221F
200062-508EA 259 USD InStock
200062-508 CA200062-508 CA200062-510 200062-510
Anti-CRYAA Rabbit Polyclonal Antibody
Antibodies
Alpha-crystallins, which are part of the small Heat shock family members, are major water-soluble proteins present in the lens of the mammalian eye. Phosphorylation of serine residues which occurs during development and in response to stress, is intimately linked with its function. Chaperone activity requires, and is modulated by, oligomerization and is limited to binding unfolded intermediates to prevent irreversible aggregation.

Type: Primary
Antigen: CRYAA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty