Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRP55 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-600-J SPA-600J ADI-SPA-600-D SPA-600F ADI-SPA-600-F SPA-600D
200062-592EA 233 USD InStock
200062-592 CA200062-594 CA200062-592 200062-594 95046-142 CA95046-142
Anti-CRP55 Rabbit Polyclonal Antibody
Antibodies
Calreticulin (CRT) is a multifunctional, multi-compartmental protein most abundant in the ER lumen. CRT contains the ER-retrieval sequence, KDEL, and has been best characterized as a soluble molecular chaperone of new or misfolded proteins and a Ca2+- binding protein. Both CRT and its membrane bound homolog, Calnexin (CNX) interact with proteins and glycoproteins that have monoglucosylated N-glycans. The CRT/CNX cycle promotes correct folding, inhibits aggregation of folding intermediates, blocks premature oligimerization, regulates ER degradation, and provides quality control by preventing incompletely folded glycoproteins from exiting to the Golgi complex.

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, and dog calreticulin. Detects a band of ~63kDa by Western blot.

Type: Primary
Antigen: CRP55
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty