Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP25 Mouse Monoclonal Antibody [clone: G3.1]

Supplier: Enzo Life Sciences
SPA-800D SPA-800F ADI-SPA-800-D ADI-SPA-800-F
200062-648EA 590 USD InStock
CA200062-646 200062-648 CA200062-648 200062-646
Anti-HSP25 Mouse Monoclonal Antibody [clone: G3.1]
Antibodies
Hsp27 is one of the most common members of the highly conserved and ubiquitously expressed family of small heat shock proteins (sHsp), which also includes alphaB-crystallin. It is characterized by a conserved C-terminal alpha-crystallin domain consisting of two anti-parallel beta-sheets that promote oligomer formation required for its primary chaperone function as inhibitor of irreversible protein aggregation. Hsp27 oligomerization is modulated by post-translational phosphorylation of Hsp27 at three serine residues, Ser15, Ser78, and Ser82, by a variety of protein kinases including MAPKAPK-3, PKAc-alpha, p70 S6K, PKD I, and PKC-delta. Hsp27 has been shown to inhibit actin polymerization by binding of unphosphorylated Hsp27 monomers to actin intermediate filaments. Anti-apoptotic functions of Hsp27 have also been identified through interactions with DAXX7, activation of Akt, and inhibition of apoptosome formation. Evidence suggests altered expression of Hsp27 is implicated in the pathogenesis of breast, ovarian, and prostate cancer.

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot, in vitro Assay, EM

Recognizes human, mouse, rat, bovine, fish, and monkey HSP27. Detects a band of ~27kDa by Western blot.

Type: Primary
Antigen: HSP25
Clonality: Monoclonal
Clone: G3.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Fish, Human, Monkey, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty