Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]

Supplier: Enzo Life Sciences
ADI-SPA-827-F SPA-827F ADI-SPA-827-D SPA-827D
200062-740EA 280 USD InStock
CA200062-740 CA200062-742 200062-740 200062-742
Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]
Antibodies
Type: Primary
Antigen: KDEL
Clonality: Monoclonal
Clone: 10C3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mammals

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty