Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DNAK Mouse Monoclonal Antibody [clone: 8E2/2]

Supplier: Enzo Life Sciences
ADI-SPA-880-D ADI-SPA-880-F ADI-SPA-880-J
95043-760EA 2320 USD InStock
95043-760 200062-782 200062-784
Anti-DNAK Mouse Monoclonal Antibody [clone: 8E2/2]
Antibodies
DnaK possesses an autophosphorylation activity and a weak 5-nucleotidase activity, cleaving the 5 phosphate groups from both ribose and deoxyribose nucleotides. The role of DnaK in ATP-dependent protein-protein interactions has been extended to normal E. coli physiology, where, like eukaryotic Hsp70 homologs, it is thought to participate in the assembly/disassembly of protein complexes.

  • For Western Blot

Recognizes bacteria DnaK. Detects a band of ~70kDa by Western blot.

Type: Primary
Antigen: DNAK
Clonality: Monoclonal
Clone: 8E2/2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty