Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDI Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-890-D SPA-890F SPA-890D ADI-SPA-890-F
200062-800EA 513 USD InStock
CA200062-800 200062-800 CA200062-798 200062-798
Anti-PDI Rabbit Polyclonal Antibody
Antibodies
The mammalian PDI (Protein disulfide-isomerase) family encompasses several highly divergent proteins which are involved in the processing and maturation of secretory proteins in the endoplasmic reticulum by catalyzing the rearrangement of disulfide bonds.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig, sheep and Xenopus PDI. Detects a band of ~58kDa by Western blot.

Type: Primary
Antigen: PDI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty