Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FKBP59 Mouse Monoclonal Antibody [clone: KN382/EC1]

Supplier: Enzo Life Sciences
ADI-SRA-1400-F SRA-1400F SRA-1400D ADI-SRA-1400-D
200062-848EA 280 USD InStock
200062-848 CA200063-288 200063-288 CA200062-848
Anti-FKBP59 Mouse Monoclonal Antibody [clone: KN382/EC1]
Antibodies
FKBP59, also known as Hsp56/p59 is an immunophilin that is a component of Hsp70/Hsp90/steroid receptor complexes. Expression of FKBP59 is increased in response to heat shock and ischemic stress, and is thought to play a role in the folding and cytoplasmic/nuclear distribution of steroid receptors.

Type: Primary
Antigen: FKBP59
Clonality: Monoclonal
Clone: KN382/EC1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Monkey, Bovine, Hamster, Mink, Rabbit, Guinea Pig

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty