Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SYT Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
SYA-148D ADI-SYA-148-D ADI-SYA-148-F SYA-148F
200062-862EA 287.58 USD InStock
CA200062-862 CA200062-864 200062-862 200062-864
Anti-SYT Mouse Monoclonal Antibody
Antibodies
Synaptotagmins (Syt) are membrane-trafficking proteins characterized by an N-terminal transmembrane region, a variable linker, and two C-terminal C2-domains, C2A and C2B. At least twelve Synaptotagamins are expressed in vertebrates and can be found in both vesicular and plasma membranes.

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes mouse, rat, fish (shark) and monkey Synaptotagmin. Detects a band of ~65kDa by Western blot.

Type: Primary
Antigen: SYT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty