Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RAP1A Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-GP125E KAP-GP125-E
200063-164EA 406 USD InStock
CA200063-164 200063-164
Anti-RAP1A Rabbit Polyclonal Antibody
Antibodies
Rap1, which is a member of the Ras family of GTP-binding proteins, cycles between an active GTP-bound and an unactive GDP-bound form that is mediated by GTPase activating protein (GAP). Rap1 is proposed to regulate Ras-mediated signaling and may also be involved in the regulation of Integrin-mediated cell adhesion although the mechanism of regulation is not known.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus Rap1. Detects a band of ~25kDa by Western blot.

Type: Primary
Antigen: RAP1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty