Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ESR1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
SRA-1010F ADI-SRA-1010-F SRA-1010D ADI-SRA-1010-D
200063-286EA 547 USD InStock
200063-286 CA200063-286 200062-846 CA200062-846
Anti-ESR1 Mouse Monoclonal Antibody
Antibodies
The Estrogen receptors (ER), mainly ERalpha and ERbeta, are members of the nuclear steroid hormone receptor family and are composed of three domains, an N-terminal domain required for maximal transactivation function (TAF), a DNA binding domain, and a steroid binding domain. Estrogen has pleiotropic effects on the growth, development, and maintenance of diverse tissues.

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat and hamster estrogen receptor alpha. Detects a band of 66.3kDa by Western blot.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty