Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NEB®Turbo Competent E.coli (High Effic.), New England Biolabs
Supplier: New England Biolabs
C2984I C2984I C2984H C2984H
200064-396EA 301 USD InStock
CA101417-896 200064-396 101417-896 CA200064-396
NEB®Turbo Competent E.coli (High Effic.), New England Biolabs
Competent Cells
Chemically competent E.coli cells suitable for high efficiency transformation and rapid colony growth (6.5 hours).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty