Would you like to log in again?
9°N™ DNA Ligase, New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M0238S M0238S
200064-420EA 90 USD InStock
CA200064-420 200064-420
9°N™ DNA Ligase, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
9°N™ DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5´ phosphate and 3´ hydroxyl termini of two adjacent oligonucleotides which are hybridized to a complementary target DNA
9°N DNA Ligase is active at elevated temperatures (45°C-90°C).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty