Would you like to log in again?
T4 PDG (T4 Endonuclease V), New England Biolabs
Supplier: New England Biolabs
M0308S M0308S
200064-436EA 85 USD InStock
CA200064-436 200064-436
T4 PDG (T4 Endonuclease V), New England Biolabs
Enzymes
T4 PDG (pyrimidine dimer glycosylase) has both DNA glycosylase and AP-lyase activity
The 16 kd protein recognizes cis-syn-cyclobutane pyrimidine dimers caused by UV irradiation. The enzyme cleaves the glycosyl bond of the 5´ end of the pyrimidine dimer and the endonucleolytic activity cleaves the phosphodiester bond at the apurinic/apyrimidinic (AP) site.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty