Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
dsRNA Ladder, New England Biolabs
Supplier: New England Biolabs
N0363S N0363S
200064-448EA 109 USD InStock
CA200064-448 200064-448
dsRNA Ladder, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
The dsRNA Ladder is a set of 7 annealed double-stranded RNA molecules produced by in vitro transcription of 14 linear DNA templates
The ladder sizes are: 500, 300, 150, 80, 50, 30 and 21 base pairs. All have 2-base, 3´ extensions. The 80 base pair fragment is at double intensity to serve as a reference point. This ladder is suitable for use as a size standard in dsRNA and RNAi analysis on both non-denaturing polyacrylamide and agarose gels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty