Would you like to log in again?
sRANKL, Total, Human ELISA, Osteoprotegerin Ligand, BioVendor
Supplier: BioVendor
RD193004200
200068-236EA 929.25 USD InStock
200068-236
sRANKL, Total, Human ELISA, Osteoprotegerin Ligand, BioVendor
Assays ELISAs
sRANKL, receptor activator of nuclear factor (NF)-κB ligand (also: osteoprotegerin ligand, OPGL), is a part of the TNF superfamily with high similarity to other members of that protein species.
  • Calibration Range: 0.5-32 pmol/mL
  • Limit of Detection: 0.4 pmol/mL
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 5 uL/well
sRANKL, receptor activator of nuclear factor (NF)-κB ligand (also: osteoprotegerin ligand, OPGL), is a part of the TNF superfamily with high similarity to other members of that protein species. Three isoforms are produced by alternate splicing, two type II membrane proteins (ISOFORM 1, 317 AA, and ISOFORM 3, 270 AA), and a secreted molecule (ISOFORM 2, 244 AA). ISOFORM 1 is identical to previously reported RANKL and possesses intracellular, transmembrane, and extracellular domains; ISOFORM 2 does not have the intracellular and transmembrane domains, and ISOFORM 3 does not have the intracellular domain. A soluble form arises by proteolytic processing from membrane isoforms.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty