Would you like to log in again?
Immunoglobulin Free Light Chains Kappa and Lambda Human ELISA, BioVendor
Supplier: BioVendor
RD194088100
200068-238EA 834.92 USD InStock
200068-238
Immunoglobulin Free Light Chains Kappa and Lambda Human ELISA, BioVendor
Assays ELISAs
In serum, FLC kappa exists predominantly as a monomer with a molecular weight of 22.5 kDa and FLC lambda as a dimer with a molecular weight of 45 kDa.
  • Calibration Range: 10–320 ug/L (FLC kappa) 17.5–560 ug/L (FLC lambda)
  • Limit of Detection: 0.6 ug/L
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum, Urine: 2x5 uL/well
Human immunoglobulin molecules consist of two identical heavy chains which define immunoglobulin classes (IgG, IgA, IgM, IgD and IgE) and identical light chains (kappa or lambda) that are covalently linked to a heavy chain. In healthy individuals, the majority of light chains in serum exist bound to heavy chain. However, low levels of free light chains (FLCs) are found in serum of normal individuals due to their excess production over heavy chains by mature B-cells. In serum, FLC kappa exists predominantly as a monomer with a molecular weight of 22.5 kDa and FLC lambda as a dimer with a molecular weight of 45 kDa. This size difference results in a differential glomerular filtration rate and, consequently, a ratio of FLC kappa to FLC lambda of 1:1.6 in serum. FLCs are observed in urine too but filtration and reabsorption of low molecular proteins in the kidney strongly affects the FLC concentration so that urinary FLC level is low in healthy individuals.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty